Learn More
Apexbio Technology LLC Ganetespib (STA-9090)(Synonyms: STA-9090, Ganetespib, STA9090, STA 9090), 10mM (in 1mL DMSO), CAS: 888216-25-9.
SDP
Catalog No. 5000060534
Encompass_Preferred
Ganetespib (STA-9090 CAS 888216-25-9) is a small-molecule inhibitor targeting heat shock protein 90 (Hsp90) It binds competitively to the ATP-binding pocket located at the N-terminal region of Hsp90 disrupting its chaperone activity and subsequently causing degradation of client proteins involved in tumor growth and survival Structurally distinct from geldanamycin-derived inhibitors Ganetespib features a triazolone moiety Preclinical studies have demonstrated antitumor activity across various cancer cell types including lung prostate colon breast cancers melanoma and leukemia highlighting its therapeutic potential for cancer research
Catalog No. 50-000-60534 Supplier Apexbio Technology LLC Supplier No. A43855.1
Ganetespib (STA-9090 CAS 888216-25-9) is a small-molecule inhibitor targeting heat shock protein 90 (Hsp90) It binds competitively to the ATP-binding pocket located at the N-terminal region of Hsp90 disrupting its chaperone activity and subsequently causing degradation of client proteins involved in tumor growth and survival Structurally distinct from geldanamycin-derived inhibitors Ganetespib features a triazolone moiety Preclinical studies have demonstrated antitumor activity across various cancer cell types including lung prostate colon breast cancers melanoma and leukemia highlighting its therapeutic potential for cancer research