Learn More
Abcam Ganetespib (STA-9090), 25MG
Catalog No. 502671872
Encompass_Preferred
MW 364.4 Da, Purity >98%. Potent Hsp90 inhibitor (IC₅₀ of 4nM in OSA 8 cells). Causes cytotoxicity in a variety of hematological and solid tumor cell lines by causing degradation of multiple oncogenic Hsp90 client proteins. Ganetespib inhibits MG63 cell lines with IC₅₀ of 43 nM.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1872 Supplier Abcam Supplier No. AB27360025MG
MW 364.4 Da, Purity >98%. Potent Hsp90 inhibitor (IC₅₀ of 4nM in OSA 8 cells). Causes cytotoxicity in a variety of hematological and solid tumor cell lines by causing degradation of multiple oncogenic Hsp90 client proteins. Ganetespib inhibits MG63 cell lines with IC₅₀ of 43 nM.
The product is subject to the following: Abcam Restricted Use Statement