Learn More
Medchemexpress LLC Ganfeborole hydrochl 100mg | 2131798-13-3 | 100MG
SDP
Catalog No. 5000241543
Encompass_Preferred
Ganfeborole hydrochloride (GSK656) is a potent antitubercular agent acting as an inhibitor of Mycobacterium tuberculosis (Mtb) leucyl-tRNA synthetase (LeuRS) with an IC50 of 0 2 M
Catalog No. 50-002-41543 Supplier Medchemexpress LLC Supplier No. HY107775100MG
Ganfeborole hydrochloride (GSK656) is a potent antitubercular agent acting as an inhibitor of Mycobacterium tuberculosis (Mtb) leucyl-tRNA synthetase (LeuRS) with an IC50 of 0 2 M