Learn More
Medchemexpress LLC Ganfeborole hydrochloride | 2131798-13-3 | 293.94 g/mol | C10H14BCl2NO4 | 25 MG
SDP
Catalog No. 5000242409
Encompass_Preferred
Ganfeborole hydrochloride is a research-grade antitubercular small molecule that inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (LeuRS) and is supplied as the hydrochloride salt for laboratory research.
  • Inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (IC50 ≈ 0.2 μM).
  • Supplied as a hydrochloride salt to facilitate handling and solubility.
  • Molecular weight 293.94 g/mol (hydrochloride).
  • Used for antitubercular research and preclinical studies.
  • Provided with safety and handling information in an SDS.
Catalog No. 50-002-42409 Supplier Medchemexpress LLC Supplier No. HY10777525MG
Ganfeborole hydrochloride is a research-grade antitubercular small molecule that inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (LeuRS) and is supplied as the hydrochloride salt for laboratory research.
  • Inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (IC50 ≈ 0.2 μM).
  • Supplied as a hydrochloride salt to facilitate handling and solubility.
  • Molecular weight 293.94 g/mol (hydrochloride).
  • Used for antitubercular research and preclinical studies.
  • Provided with safety and handling information in an SDS.