Learn More
Medchemexpress LLC Ganfeborole hydrochloride | 2131798-13-3 | MFCD31657361 | 100.0% | 293.94 | C10H14BCl2NO4 | 1 MG
SDP
Catalog No. 5000241544
Encompass_Preferred
Ganfeborole hydrochloride is a small-molecule antitubercular agent that inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (LeuRS). Supplied as an off-white to light-yellow solid, it is intended for research use in biochemical and cellular assays; reported IC50 against Mtb LeuRS is 0.2 μM. Store sealed and away from moisture; in solvent, store at -80°C for long-term stability.
  • LeuRS inhibitor with reported IC50 0.2 μM.
  • High purity suitable for biochemical assays.
  • Off-white to light-yellow solid form, convenient for handling.
  • Available in multiple small-scale quantities for research workflows.
  • Storage guidance provided for solid and solution stability.
Catalog No. 50-002-41544 Supplier Medchemexpress LLC Supplier No. HY1077751MG
Ganfeborole hydrochloride is a small-molecule antitubercular agent that inhibits Mycobacterium tuberculosis leucyl-tRNA synthetase (LeuRS). Supplied as an off-white to light-yellow solid, it is intended for research use in biochemical and cellular assays; reported IC50 against Mtb LeuRS is 0.2 μM. Store sealed and away from moisture; in solvent, store at -80°C for long-term stability.
  • LeuRS inhibitor with reported IC50 0.2 μM.
  • High purity suitable for biochemical assays.
  • Off-white to light-yellow solid form, convenient for handling.
  • Available in multiple small-scale quantities for research workflows.
  • Storage guidance provided for solid and solution stability.