Learn More
AdipoGen Ganglioside GD1b 2Na bovine
SDP
Catalog No. 502562468
Encompass_Preferred
Chemical. CAS 19553-76-5. Formula C84H146N4O39 . 2Na. MW 1836.1 . 46.0 calculated on sphingosine C181 and stearic acid. Isolated from bovine brain. Gangliosides are acidic glycosphingolipids that form lipid rafts in the outer leaflet of the cell plasma membrane, especially in neuronal cells in the central nervous system. They participate in cellular proliferation, differentiation, adhesion, signal transduction, cell-to-cell interactions, tumorigenesis and metastasis. The accumulation of gangliosides has been linked to several diseases. Ganglioside GD1b may act as a bacterial toxin receptor tetanus, botulinus. High levels are found in gliomas and astrocytomas. It modulates protein kinase activities. In vivo and in vitro it is in equilibrium with the lactone structure.
Catalog No. 50-256-2468 Supplier AdipoGen Supplier No. AGCN29004M001
Chemical. CAS 19553-76-5. Formula C84H146N4O39 . 2Na. MW 1836.1 . 46.0 calculated on sphingosine C181 and stearic acid. Isolated from bovine brain. Gangliosides are acidic glycosphingolipids that form lipid rafts in the outer leaflet of the cell plasma membrane, especially in neuronal cells in the central nervous system. They participate in cellular proliferation, differentiation, adhesion, signal transduction, cell-to-cell interactions, tumorigenesis and metastasis. The accumulation of gangliosides has been linked to several diseases. Ganglioside GD1b may act as a bacterial toxin receptor tetanus, botulinus. High levels are found in gliomas and astrocytomas. It modulates protein kinase activities. In vivo and in vitro it is in equilibrium with the lactone structure.