Learn More
Medchemexpress LLC Ganglioside GQ1b (bovine) sodium | 68652-37-9 | >98.0% | 2508.5 g/mol | C106H178N6O55Na4 | 1mg
SDP
Catalog No. 5000270292
Encompass_Preferred
Ganglioside GQ1b (bovine) sodium promotes neurite outgrowth during early neuronal differentiation Ganglioside GQ1b (bovine) sodium controls the neuronal differentiation process in mESCs[1]
Catalog No. 50-002-70292 Supplier Medchemexpress LLC Supplier No. HYW7475991MG
Ganglioside GQ1b (bovine) sodium promotes neurite outgrowth during early neuronal differentiation Ganglioside GQ1b (bovine) sodium controls the neuronal differentiation process in mESCs[1]