Learn More
Bioworld GANIRELIX
Supplier:  Bioworld 220600811
Encompass_Preferred
Ganirelix Description: Ganirelix acetate is a synthetic decapeptide with high antagonistic activity against naturally occurring gonadotropin-releasing hormone (GnRH). Ganirelix acetate is derived from native GnRH with substitutions of ...
Catalog No. 50-253-178