Learn More
Medchemexpress LLC Ganirelix acetate | 129311-55-3 | 99.19% | 1690.42 | 1 ML
SDP
Catalog No. 5000343730
Encompass_Preferred
Ganirelix acetate is an injectable competitive gonadotropin-releasing hormone (GnRH) antagonist. It directly competes against the endogenous molecule for receptor binding, leading to a rapid reduction in estradiol levels. It can be used for researching ovarian hyperstimulation syndrome (OHSS).
  • Targets GnRH with an IC50 of 3.6 nM in HEK293 cells
  • Antagonizes human GnHR receptor activity
  • Suitable for research on ovarian hyperstimulation syndrome (OHSS)
  • Causes rapid reduction in estradiol levels
Catalog No. 50-003-43730 Supplier Medchemexpress LLC Supplier No. HY10953210MG
Ganirelix acetate is an injectable competitive gonadotropin-releasing hormone (GnRH) antagonist. It directly competes against the endogenous molecule for receptor binding, leading to a rapid reduction in estradiol levels. It can be used for researching ovarian hyperstimulation syndrome (OHSS).
  • Targets GnRH with an IC50 of 3.6 nM in HEK293 cells
  • Antagonizes human GnHR receptor activity
  • Suitable for research on ovarian hyperstimulation syndrome (OHSS)
  • Causes rapid reduction in estradiol levels