Learn More
Medchemexpress LLC Ganirelix acetate | 129311-55-3 | 99.19% | 1690.42 | 50 MG
SDP
Catalog No. 5000343397
Encompass_Preferred
Ganirelix acetate (Ganirest) is an injectable competitive gonadotropin-releasing hormone (GnRH) antagonist. It directly competes against the endogenous molecule for receptor binding, leading to a rapid reduction in estradiol levels. Ganirelix acetate can be used for researching ovarian hyperstimulation syndrome (OHSS).
  • Injectable competitive gonadotropin-releasing hormone (GnRH) antagonist.
  • Directly competes against endogenous molecules for receptor binding.
  • Causes a rapid reduction in estradiol levels.
  • Can be used for researching ovarian hyperstimulation syndrome (OHSS).
  • For research use only.
Catalog No. 50-003-43397 Supplier Medchemexpress LLC Supplier No. HY1095321MG
Ganirelix acetate (Ganirest) is an injectable competitive gonadotropin-releasing hormone (GnRH) antagonist. It directly competes against the endogenous molecule for receptor binding, leading to a rapid reduction in estradiol levels. Ganirelix acetate can be used for researching ovarian hyperstimulation syndrome (OHSS).
  • Injectable competitive gonadotropin-releasing hormone (GnRH) antagonist.
  • Directly competes against endogenous molecules for receptor binding.
  • Causes a rapid reduction in estradiol levels.
  • Can be used for researching ovarian hyperstimulation syndrome (OHSS).
  • For research use only.