Learn More
Medchemexpress LLC Ganirelix acetate | 129311-55-3 | MFCD00026356 | 99.2% | 1690.4 g/mol | C84H121ClN18O17 | 5 MG
SDP
Catalog No. 5000263341
Encompass_Preferred
Ganirelix acetate is a synthetic gonadotropin-releasing hormone (GnRH) receptor antagonist provided as a research-grade peptide. It is used to suppress endogenous GnRH signaling in studies of reproductive biology, ovarian stimulation protocols, and related pharmacology. For research use only.
  • Inhibits GnRH receptor activity, reducing luteinizing hormone release.
  • Suitable for in vitro and in vivo reproductive research.
  • Supplied as lyophilized powder for convenient reconstitution.
  • High purity appropriate for analytical and biological assays.
  • Accompanied by certificate of analysis and batch-specific data.
Catalog No. 50-002-63341 Supplier Medchemexpress LLC Supplier No. HY1095325MG
Ganirelix acetate is a synthetic gonadotropin-releasing hormone (GnRH) receptor antagonist provided as a research-grade peptide. It is used to suppress endogenous GnRH signaling in studies of reproductive biology, ovarian stimulation protocols, and related pharmacology. For research use only.
  • Inhibits GnRH receptor activity, reducing luteinizing hormone release.
  • Suitable for in vitro and in vivo reproductive research.
  • Supplied as lyophilized powder for convenient reconstitution.
  • High purity appropriate for analytical and biological assays.
  • Accompanied by certificate of analysis and batch-specific data.