Learn More
Electron Microscopy Sciences EMS Gannetts PM Dissecting Knife, 10" OAL
SDP
Catalog No. 501931582
Encompass_Preferred
The broad non-flexible blade features an extra thick cutting edge perfect for cutting dense tissue. The top edge can be struck with a mallet to hammer through hard tissue and/or bone. Made of heavy duty stainless steel.
OAL: 10"
Catalog No. 50-193-1582 Supplier Electron Microscopy Sciences Supplier No. 7205901
The broad non-flexible blade features an extra thick cutting edge perfect for cutting dense tissue. The top edge can be struck with a mallet to hammer through hard tissue and/or bone. Made of heavy duty stainless steel.
OAL: 10"