Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ganoderal A | 104700-98-3 | 97.5% | 436.67 | C30H44O2 | 10 MG
SDP

Catalog No. 5000295697
Encompass_Preferred

Ganoderal A is an oxygenated sterol isolated from Ganoderma lucidum that acts as a cholesterol synthesis inhibitor. It is provided as a white to off-white solid for research use, with reported purity of 97.5% and molecular weight of 436.67.

  • White to off-white solid suitable for analytical use.
  • Reported purity 97.5%.
  • CAS number 104700-98-3 for unambiguous identification.
  • Molecular formula C30H44O2 and molecular weight 436.67.
  • Supplied in a 10 mg quantity for laboratory research.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-95697 Supplier Medchemexpress LLC Supplier No. HYN222110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ganoderal A is an oxygenated sterol isolated from Ganoderma lucidum that acts as a cholesterol synthesis inhibitor. It is provided as a white to off-white solid for research use, with reported purity of 97.5% and molecular weight of 436.67.

  • White to off-white solid suitable for analytical use.
  • Reported purity 97.5%.
  • CAS number 104700-98-3 for unambiguous identification.
  • Molecular formula C30H44O2 and molecular weight 436.67.
  • Supplied in a 10 mg quantity for laboratory research.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.