Learn More
Medchemexpress LLC Ganoderenic acid A | 100665-40-5 | MFCD07779161 | 97.9% | 514.65 | C30H42O7 | 10 MG
SDP
Catalog No. 5000289134
Encompass_Preferred
Ganoderenic acid A is a lanostane-type triterpene isolated from Ganoderma species used in biochemical research as a tool compound and inhibitor of β-glucuronidase. It is provided in solid and solution formats for enzymatic and cellular assays.
  • Inhibitor of β-glucuronidase, useful for enzymatic and metabolic studies.
  • Available as a 10 mg solid and as 10 mM solutions in DMSO for assay preparation.
  • Characterized by CAS 100665-40-5, formula C30H42O7, molecular weight 514.65.
  • Quality-tested with HPLC purity approximately 97.9% as reported on the certificate of analysis.
Catalog No. 50-002-89134 Supplier Medchemexpress LLC Supplier No. HYN299810MG
Ganoderenic acid A is a lanostane-type triterpene isolated from Ganoderma species used in biochemical research as a tool compound and inhibitor of β-glucuronidase. It is provided in solid and solution formats for enzymatic and cellular assays.
  • Inhibitor of β-glucuronidase, useful for enzymatic and metabolic studies.
  • Available as a 10 mg solid and as 10 mM solutions in DMSO for assay preparation.
  • Characterized by CAS 100665-40-5, formula C30H42O7, molecular weight 514.65.
  • Quality-tested with HPLC purity approximately 97.9% as reported on the certificate of analysis.