Learn More
TARGETMOL CHEMICALS INC Ganoderenic acid A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN16565MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes.Ganoderenic acid A has a potent hepatoprotective, and cytotoxic effects, it shows inhibitory activity on human aldose reductase in vitro. Purity 98.5%
Catalog No. 50-288-0263