Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ganoderenic acid E | 110241-23-1 | 1 MG
SDP

Catalog No. 5000418521
Encompass_Preferred

Ganoderenic acid E is a lanostane-type triterpene with a purity of 95.0%. It has a molecular weight of 528.63 and a molecular formula of C30H40O8. This compound appears as a white to off-white solid.

  • Lanostane-type triterpene
  • Purity of 95.0%
  • Molecular weight: 528.63
  • Molecular formula: C30H40O8
  • Appearance: solid
  • Color: white to off-white

Catalog No. 50-004-18521 Supplier Medchemexpress LLC Supplier No. HYN68671MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ganoderenic acid E is a lanostane-type triterpene with a purity of 95.0%. It has a molecular weight of 528.63 and a molecular formula of C30H40O8. This compound appears as a white to off-white solid.

  • Lanostane-type triterpene
  • Purity of 95.0%
  • Molecular weight: 528.63
  • Molecular formula: C30H40O8
  • Appearance: solid
  • Color: white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.