Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ganoderic acid A | 81907-62-2 | 516.67 | 1 ML
SDP

Catalog No. 5000414666
Encompass_Preferred

Ganoderic acid A can inhibit the JAK-STAT3 signaling pathway, and also inhibit proliferation, viability, and ROS. This compound is intended for research use only and not for sale to patients.

  • Inhibits JAK-STAT3 signaling pathway
  • Inhibits proliferation, viability, and ROS
  • Cytotoxic against Bel-7402 cells with an IC50 of 7.25 μM
  • Cytotoxic against HepG2 cells with a CC50 of 80 μM and IC50 of 38.7 μM
  • Enhances HLA class II-mediated antigen presentation
  • Promotes cisplatin-induced cell death via STAT3 suppression
  • Inhibits proliferation and induces apoptosis in human osteosarcoma HOS and MG-63 cells
  • High purity of 99.26%
  • Soluble in DMSO and ethanol
  • Shipped at room temperature
  • Storage at 4°C (solid) and -80°C/-20°C (in solvent)

Catalog No. 50-004-14666 Supplier Medchemexpress LLC Supplier No. HYN144710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ganoderic acid A can inhibit the JAK-STAT3 signaling pathway, and also inhibit proliferation, viability, and ROS. This compound is intended for research use only and not for sale to patients.

  • Inhibits JAK-STAT3 signaling pathway
  • Inhibits proliferation, viability, and ROS
  • Cytotoxic against Bel-7402 cells with an IC50 of 7.25 μM
  • Cytotoxic against HepG2 cells with a CC50 of 80 μM and IC50 of 38.7 μM
  • Enhances HLA class II-mediated antigen presentation
  • Promotes cisplatin-induced cell death via STAT3 suppression
  • Inhibits proliferation and induces apoptosis in human osteosarcoma HOS and MG-63 cells
  • High purity of 99.26%
  • Soluble in DMSO and ethanol
  • Shipped at room temperature
  • Storage at 4°C (solid) and -80°C/-20°C (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.