Learn More
Medchemexpress LLC Ganoderic acid C1 | 95311-97-0 | 95.0% | 1 MG
SDP
Catalog No. 5000338373
Encompass_Preferred
Ganoderic acid C1 is a natural compound isolated from *G. lucidum*. It suppresses TNF-α production by murine macrophages (RAW 264.7 cells).
  • Isolated from *G. lucidum*
  • Suppresses TNF-α production
  • Targets TNF receptor
  • Inhibits apoptosis pathway
Catalog No. 50-003-38373 Supplier Medchemexpress LLC Supplier No. HY1291511MG
Ganoderic acid C1 is a natural compound isolated from *G. lucidum*. It suppresses TNF-α production by murine macrophages (RAW 264.7 cells).
  • Isolated from *G. lucidum*
  • Suppresses TNF-α production
  • Targets TNF receptor
  • Inhibits apoptosis pathway