Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ganoderic acid C2 | 103773-62-2 | 518.68 | 1 ML
SDP

Catalog No. 5000416005
Encompass_Preferred

Ganoderic acid C2 is a bioactive triterpenoid found in *Ganoderma lucidum*. It possesses potential anti-tumor bioactivity, antihistamine, anti-aging, and cytotoxic effects. It is an inhibitor for aldose reductase with an IC50 of 43.8 μM.

  • Purity: 99.72%
  • Bioactive triterpenoid found in *Ganoderma lucidum*
  • Potential activities include anti-tumor, antihistamine, anti-aging, and cytotoxic effects
  • Aldose reductase inhibitor with an IC50 of 43.8 μM
  • Appearance: solid, white to off-white
  • Immunomodulatory activity exhibited in Cyclophosphamide-induced immunosuppression mouse models

Catalog No. 50-004-16005 Supplier Medchemexpress LLC Supplier No. HYN151710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ganoderic acid C2 is a bioactive triterpenoid found in *Ganoderma lucidum*. It possesses potential anti-tumor bioactivity, antihistamine, anti-aging, and cytotoxic effects. It is an inhibitor for aldose reductase with an IC50 of 43.8 μM.

  • Purity: 99.72%
  • Bioactive triterpenoid found in *Ganoderma lucidum*
  • Potential activities include anti-tumor, antihistamine, anti-aging, and cytotoxic effects
  • Aldose reductase inhibitor with an IC50 of 43.8 μM
  • Appearance: solid, white to off-white
  • Immunomodulatory activity exhibited in Cyclophosphamide-induced immunosuppression mouse models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.