Learn More
Medchemexpress LLC Ganoderic acid C2 | 103773-62-2 | MFCD04039842 | 99.7% | 518.68 g/mol | C30H46O7 | 10 MG
SDP
Catalog No. 5000295585
Encompass_Preferred
Ganoderic acid C2 is a bioactive triterpenoid (C30H46O7; 518.68 g/mol; CAS 103773-62-2) isolated from Ganoderma lucidum. It has reported aldose reductase inhibitory activity and shows potential anti-tumor, antihistamine, anti-aging, and cytotoxic effects. The compound is supplied as a purified solid and is used in biochemical and cell-based research applications.
  • High purity suitable for biochemical assays (reported purity 99.7%).
  • Available as a solid and as a DMSO solution for experimental flexibility.
  • Storage-stable when handled per recommendations (4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month).
  • Useful as an aldose reductase inhibitor and in cytotoxicity and mechanism-of-action studies.
Catalog No. 50-002-95585 Supplier Medchemexpress LLC Supplier No. HYN151710MG
Ganoderic acid C2 is a bioactive triterpenoid (C30H46O7; 518.68 g/mol; CAS 103773-62-2) isolated from Ganoderma lucidum. It has reported aldose reductase inhibitory activity and shows potential anti-tumor, antihistamine, anti-aging, and cytotoxic effects. The compound is supplied as a purified solid and is used in biochemical and cell-based research applications.
  • High purity suitable for biochemical assays (reported purity 99.7%).
  • Available as a solid and as a DMSO solution for experimental flexibility.
  • Storage-stable when handled per recommendations (4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month).
  • Useful as an aldose reductase inhibitor and in cytotoxicity and mechanism-of-action studies.