Learn More
TARGETMOL CHEMICALS INC GANODERIC ACID D 10MG
SDP
Catalog No. 502576640
Encompass_Preferred
Also available in 1 mg 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Ganoderic acid D is one of the major components in Ganoderma triterpenes. 2. Ganoderic acid D treatment for 48h inhibits the proliferation of HeLa human cervical carcinoma cells with an IC(5) value of 17.3 +/- .3 microM. purity: 99%
Catalog No. 50-257-6640 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S113310MG
Also available in 1 mg 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Ganoderic acid D is one of the major components in Ganoderma triterpenes. 2. Ganoderic acid D treatment for 48h inhibits the proliferation of HeLa human cervical carcinoma cells with an IC(5) value of 17.3 +/- .3 microM. purity: 99%