Learn More
Medchemexpress LLC HY-N1511 1mg Medchemexpress, Ganoderic acid D CAS:108340-60-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN15111MG
Encompass_Preferred
Medchemexpress, HY-N1511 1mg Ganoderic acid D CAS:108340-60-9 Ganoderic acid D, a highly oxygenated tetracyclic triterpenoid, is the major active component of Ganoderma lucidum. Ganoderic acid D upregulates the protein expression of SIRT3 and induces the deacetylated cyclophilin D (CypD) by SIRT3. Ganoderic acid D inhibits the energy reprogramming of colon cancer cells including glucose uptake, lactate production, pyruvate and acetyl-coenzyme production in colon cancer cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3850