Learn More
TARGETMOL CHEMICALS INC Ganoderic acid H 1MG
SDP
Catalog No. 502879384
Encompass_Preferred
Also available in 5 mg and bulk. Please contact Fisher for quotes. Ganoderic acid H is a potent antitumour agent, it mediates its biological effects through the inhibition of transcription factors AP-1 and NF-kB, resulting in the suppression of secretion of uPA. Purity 95%
Catalog No. 50-287-9384 Supplier TARGETMOL CHEMICALS INC Supplier No. TN16641MG
Also available in 5 mg and bulk. Please contact Fisher for quotes. Ganoderic acid H is a potent antitumour agent, it mediates its biological effects through the inhibition of transcription factors AP-1 and NF-kB, resulting in the suppression of secretion of uPA. Purity 95%