Learn More
Medchemexpress LLC HY-N1513 5mg Medchemexpress, Ganoderic acid H CAS:98665-19-1 Purity:>98%
SDP
Catalog No. 502082848
Encompass_Preferred
Medchemexpress, HY-N1513 5mg Ganoderic acid H CAS:98665-19-1 Ganoderic acid H is a lanostane-type triterpene isolated from Ganoderma lucidum. Ganoderic acid H suppresses growth and invasive behavior of breast cancer cells through the inhibition of transcription factors AP-1 and NF-kappaB signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2848 Supplier Medchemexpress LLC Supplier No. HYN15135MG
Medchemexpress, HY-N1513 5mg Ganoderic acid H CAS:98665-19-1 Ganoderic acid H is a lanostane-type triterpene isolated from Ganoderma lucidum. Ganoderic acid H suppresses growth and invasive behavior of breast cancer cells through the inhibition of transcription factors AP-1 and NF-kappaB signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.