Learn More
Medchemexpress LLC Ganoderic acid T-Q | 112430-66-7 | 5 MG
SDP
Catalog No. 5000419833
Encompass_Preferred
Ganoderic acid T-Q is a natural product found in Ganoderma lucidum that stimulates tubulin polymerization. This product is a white to off-white solid with high purity, intended for research applications only.
  • Stimulates tubulin polymerization
  • Natural product from Ganoderma lucidum
  • White to off-white solid appearance
  • Purity of 99.41%
  • Intended for research use only
Catalog No. 50-004-19833 Supplier Medchemexpress LLC Supplier No. HYN80395MG
Ganoderic acid T-Q is a natural product found in Ganoderma lucidum that stimulates tubulin polymerization. This product is a white to off-white solid with high purity, intended for research applications only.
  • Stimulates tubulin polymerization
  • Natural product from Ganoderma lucidum
  • White to off-white solid appearance
  • Purity of 99.41%
  • Intended for research use only