Learn More
Medchemexpress LLC Ganoderic acid T-Q | 112430-66-7 | MFCD21333562 | 1 MG
SDP
Catalog No. 5000419369
Encompass_Preferred
Ganoderic acid T-Q is a natural product isolated from *Ganoderma lucidum*. This compound has been identified as stimulating tubulin polymerization. It is intended for research use only.
  • Natural product source
  • Stimulates tubulin polymerization
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-19369 Supplier Medchemexpress LLC Supplier No. HYN80391MG
Ganoderic acid T-Q is a natural product isolated from *Ganoderma lucidum*. This compound has been identified as stimulating tubulin polymerization. It is intended for research use only.
  • Natural product source
  • Stimulates tubulin polymerization
  • Solid appearance
  • White to off-white color