Learn More
Medchemexpress LLC Ganodermanontriol | 106518-63-2 | 472.70 | 1 MG
SDP
Catalog No. 5000414774
Encompass_Preferred
Ganodermanontriol is a sterol isolated from *Ganoderma lucidum* that induces anti-inflammatory activity in tert-butyl hydroperoxide (t-BHP)-damaged hepatic cells through the expression of HO-1. It also exhibits hepatoprotective activity and antiproliferative activity against human MCF7 and MDA-MB-231 breast cancer cells.
  • Induces anti-inflammatory activity.
  • Exhibits hepatoprotective activity.
  • Suppresses growth of colon cancer cells through β-catenin signaling.
  • Exhibits anti-melanogenic effects through regulation of CREB and MAPK signaling pathways.
  • Inhibits breast cancer cell lines MCF-7 and MDA-MB-231.
Catalog No. 50-004-14774 Supplier Medchemexpress LLC Supplier No. HYN15061MG
Ganodermanontriol is a sterol isolated from *Ganoderma lucidum* that induces anti-inflammatory activity in tert-butyl hydroperoxide (t-BHP)-damaged hepatic cells through the expression of HO-1. It also exhibits hepatoprotective activity and antiproliferative activity against human MCF7 and MDA-MB-231 breast cancer cells.
  • Induces anti-inflammatory activity.
  • Exhibits hepatoprotective activity.
  • Suppresses growth of colon cancer cells through β-catenin signaling.
  • Exhibits anti-melanogenic effects through regulation of CREB and MAPK signaling pathways.
  • Inhibits breast cancer cell lines MCF-7 and MDA-MB-231.