Learn More
Medchemexpress LLC Ganodermanontriol | 106518-63-2 | 472.70 | 5 MG
SDP
Catalog No. 5000415048
Encompass_Preferred
Ganodermanontriol is a sterol isolated from Ganoderma lucidum that induces anti-inflammatory activity in tert-butyl hydroperoxide (t-BHP)-damaged hepatic cells by expressing HO-1. It also exhibits hepatoprotective activity. It has been shown to inhibit breast cancer cell lines MCF-7 and MDA-MB-231 with IC50 values of 5.8 and 9.7 μM, respectively.
  • Induces anti-inflammatory activity in damaged hepatic cells.
  • Exhibits hepatoprotective activity.
  • Inhibits breast cancer cell lines MCF-7 and MDA-MB-231.
Catalog No. 50-004-15048 Supplier Medchemexpress LLC Supplier No. HYN15065MG
Ganodermanontriol is a sterol isolated from Ganoderma lucidum that induces anti-inflammatory activity in tert-butyl hydroperoxide (t-BHP)-damaged hepatic cells by expressing HO-1. It also exhibits hepatoprotective activity. It has been shown to inhibit breast cancer cell lines MCF-7 and MDA-MB-231 with IC50 values of 5.8 and 9.7 μM, respectively.
  • Induces anti-inflammatory activity in damaged hepatic cells.
  • Exhibits hepatoprotective activity.
  • Inhibits breast cancer cell lines MCF-7 and MDA-MB-231.