Learn More
Medchemexpress LLC Ganoderol A | 104700-97-2 | 1 MG
SDP
Catalog No. 5000420147
Encompass_Preferred
Ganoderol A is a terpenoid extracted from *Ganoderma lucidum* with antimicrobial activities. It inhibits the cholesterol synthesis pathway and exhibits significant anti-inflammatory activity and protection against ultraviolet A (UVA) damage.
  • Antimicrobial activities
  • Inhibits cholesterol synthesis pathway
  • Significant anti-inflammatory activity
  • Protection against ultraviolet A (UVA) damage
Catalog No. 50-004-20147 Supplier Medchemexpress LLC Supplier No. HYN39251MG
Ganoderol A is a terpenoid extracted from *Ganoderma lucidum* with antimicrobial activities. It inhibits the cholesterol synthesis pathway and exhibits significant anti-inflammatory activity and protection against ultraviolet A (UVA) damage.
  • Antimicrobial activities
  • Inhibits cholesterol synthesis pathway
  • Significant anti-inflammatory activity
  • Protection against ultraviolet A (UVA) damage