Learn More
Medchemexpress LLC HY-13282 5mg Medchemexpress, GANT 58 CAS:64048-12-0 Purity:>98%
SDP
Catalog No. 502081323
Encompass_Preferred
Medchemexpress, HY-13282 5mg GANT 58 CAS:64048-12-0 GANT 58 is a potent Gli antagonist that inhibits GLI1-induced transcription with IC50 of 5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1323 Supplier Medchemexpress LLC Supplier No. HY132825MG
Medchemexpress, HY-13282 5mg GANT 58 CAS:64048-12-0 GANT 58 is a potent Gli antagonist that inhibits GLI1-induced transcription with IC50 of 5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.