Learn More
Medchemexpress LLC HY-13901 10mg , GANT 61 CAS:500579-04-4 Purity:>98%
SDP
Catalog No. 502259725
Encompass_Preferred
HY-13901 10mg GANT 61 CAS: 500579-04-4 GANT 61 is an inhibitor of Gli1 and Gli2 targeting the Hedgehog/GLI pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9725 Supplier Medchemexpress LLC Supplier No. HY1390110MG
HY-13901 10mg GANT 61 CAS: 500579-04-4 GANT 61 is an inhibitor of Gli1 and Gli2 targeting the Hedgehog/GLI pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.