Learn More
Abcam GANT 61, GLI1 and 2 inhibitor, 10MG
Catalog No. 502707144
Encompass_Preferred
MW 429.61 g/mol, Purity >98%. Selective GLI1 and 2 inhibitor (IC₅₀ = 5 μM). Blocks the hedgehog/smoothened signaling pathway. Anticancer activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7144 Supplier Abcam Supplier No. AB12090410MG
MW 429.61 g/mol, Purity >98%. Selective GLI1 and 2 inhibitor (IC₅₀ = 5 μM). Blocks the hedgehog/smoothened signaling pathway. Anticancer activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement