Learn More
Abcam GANT58, Hh signaling inhibitor, 10MG
Catalog No. 502707370
Encompass_Preferred
MW 392.48 g/mol, Purity >99%. Inhibits Hh signaling downstream of smo and sufu. IC₅₀ = 5 μM for glioma associated oncogene expression in MEFs.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7370 Supplier Abcam Supplier No. AB12066510MG
MW 392.48 g/mol, Purity >99%. Inhibits Hh signaling downstream of smo and sufu. IC₅₀ = 5 μM for glioma associated oncogene expression in MEFs.
The product is subject to the following: Abcam Restricted Use Statement