Learn More
Selleck Chemical LLC GANT61 10mg 500579-04-4 NSC 136476
SDP
Catalog No. NC1366195
Encompass_Preferred
GANT61 (NSC 136476) is an inhibitor for GLI1 as well as GLI2-induced transcription, inhibits hedgehog with IC50 of 5 ?M in GLI1 expressing HEK293T cell, displays selectivity over other pathways, such as TNF and glucocorticoid receptor gene transactivation. GANT61 induces apoptosis and activates protective autophagy in LX-2 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1366195 Supplier Selleck Chemical LLC Supplier No. S807510MG
GANT61 (NSC 136476) is an inhibitor for GLI1 as well as GLI2-induced transcription, inhibits hedgehog with IC50 of 5 ?M in GLI1 expressing HEK293T cell, displays selectivity over other pathways, such as TNF and glucocorticoid receptor gene transactivation. GANT61 induces apoptosis and activates protective autophagy in LX-2 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.