Learn More
AdipoGen GANT61 (1 mg)
SDP
Supplier:  AdipoGen AGCR13561M001
Encompass_Preferred
Chemical. CAS: 500579-04-4. Formula: C27H35N5. MW: 429.6. Cell permeable GLI antagonist. Inhibits Gli1 and Gli2-induced transcription. Sonic hedgehog (Shh) signaling pathway inhibitor, downstream of SMO and SUFU. Displays antiproliferative and antitumor activity in vitro and in vivo. Apoptosis inducer.
Catalog No. 50-205-3928