Learn More
AdipoGen GANT61,Chemical. CAS: 500579-04-4
SDP
Catalog No. 501176090
Encompass_Preferred
GANT61, AdipoGen Life Sciences. Chemical. CAS: 500579-04-4. Formula: C27H35N5. MW: 429.6. Cell permeable GLI antagonist. Inhibits Gli1 and Gli2-induced transcription. Sonic hedgehog (Shh) signaling pathway inhibitor, downstream of SMO and SUFU. Displays antiproliferative and antitumor activity in vitro and in vivo. Apoptosis inducer. 500579-04-4
Catalog No. 50-117-6090 Supplier AdipoGen Supplier No. AGCR13561M005
GANT61, AdipoGen Life Sciences. Chemical. CAS: 500579-04-4. Formula: C27H35N5. MW: 429.6. Cell permeable GLI antagonist. Inhibits Gli1 and Gli2-induced transcription. Sonic hedgehog (Shh) signaling pathway inhibitor, downstream of SMO and SUFU. Displays antiproliferative and antitumor activity in vitro and in vivo. Apoptosis inducer. 500579-04-4