Learn More
Apexbio Technology LLC Gap 26, 10mg. Cas: 197250-15-0 MFCD: MFCD06411601. Gap junction blocker peptide, mapping to connexin 43 residue 63-75
SDP
Supplier:  Apexbio Technology LLC A104410
Encompass_Preferred
Gap26 (Val-Cys-Tyr-Asp-Lys-Ser-Phe-Pro-Ile-Ser-His-Val-Arg) is a connexin mimetic peptide, corresponding to residues 63-75 of connexin 43, which is a gap junction blocker.Connexins, or gap junctions, are a family of structurally-related transmembrane proteins. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4681