Learn More
Medchemexpress LLC HY-P1082A 1mg Medchemexpress, Gap 26 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1082A1MG
Encompass_Preferred
Medchemexpress, HY-P1082A 1mg Gap 26 (TFA) CAS: Gap 26 TFA is a connexin mimetic peptide, composed of residue numbers 63-75 of the first extracellular loop of connexin 43 (gap junction blocker), containing the SHVR amino acid motif. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3038