Learn More
Cayman Chemical Gap 27 (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502911066
Encompass_Preferred
A connexin-mimetic peptide; reduces survival of isolated rat osteoclasts at 500 µM; decreases bone resorption in rat bone slice and osteoclast co-cultures; accelerates wound closure and stratification of the corneal epithelium in isolated human corneas; reduces cardiac remodeling and the incidence of cardiac arrhythmias in a rat model of AV-shunt surgery-induced high-output heart failure
Catalog No. 50-291-1066 Supplier Cayman Chemical Supplier No. 351245 MG
A connexin-mimetic peptide; reduces survival of isolated rat osteoclasts at 500 µM; decreases bone resorption in rat bone slice and osteoclast co-cultures; accelerates wound closure and stratification of the corneal epithelium in isolated human corneas; reduces cardiac remodeling and the incidence of cardiac arrhythmias in a rat model of AV-shunt surgery-induced high-output heart failure