Learn More
Medchemexpress LLC HY-P1136A 1mg Medchemexpress, Gap19 (TFA) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1136A1MG
Encompass_Preferred
Medchemexpress, HY-P1136A 1mg Gap19 (TFA) CAS:0 Gap19 TFA, a peptide derived from nine amino acids of the Cx43 cytoplasmic loop (CL), is a potent and selective connexin 43 (Cx43) hemichannel blocker. Gap19 TFA inhibits hemichannels caused by preventing intramolecular interactions of the C-terminus (CT) with the CL. Gap19 TFA is not blocking GJ channels or Cx40/pannexin-1 hemichannels. Gap19 TFA has protective effects against myocardial[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1670