Learn More
Aviva Systems Biology Corp GAP43 Antibody - C-terminal region (ARP61317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAP43. It was validated on Western Blot. . Alternate names: B-50, PP46ApplicationWB. Species reactivity: Cow, Dog, Goat, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-341