Learn More
Sigma Aldrich Fine Chemicals Biosciences GAPDH Activity Assay Kit s
Catalog No. 502904359
Encompass
GAPDH (Glyceraldehyde-3-Phosphate Dehydrogenase EC 1.2.1.12) catalyzes the conversion of Glyceraldehyde-3-Phosphate (GAP) to 1 3-Bisphosphate Glycerate (BPG) and plays a key role in glycolysis. The enzyme is involved in cellular processes such as apoptosis membrane trafficking iron metabolism and nuclear translocation. GAPDH (housekeeping gene) expression is stable and constitutive. Deregulation of GAPDH activity is associated with abnormal cell proliferation and carcinogenesis. Accurate quantitation of GAPDH activity is important for diagnosing diseases and studying normal cellular physiology.
Catalog No. 50-290-4359 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK2771KT
GAPDH (Glyceraldehyde-3-Phosphate Dehydrogenase EC 1.2.1.12) catalyzes the conversion of Glyceraldehyde-3-Phosphate (GAP) to 1 3-Bisphosphate Glycerate (BPG) and plays a key role in glycolysis. The enzyme is involved in cellular processes such as apoptosis membrane trafficking iron metabolism and nuclear translocation. GAPDH (housekeeping gene) expression is stable and constitutive. Deregulation of GAPDH activity is associated with abnormal cell proliferation and carcinogenesis. Accurate quantitation of GAPDH activity is important for diagnosing diseases and studying normal cellular physiology.