Learn More
Carbosynth LLC. Mouse monoclonal GAPDH antibody
SDP
Supplier:  Carbosynth LLC. 10RG109A1MG
Encompass_Preferred
Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) is an enzyme that catalyzes the sixth step of glycolysis and thus serves to break down glucose for energy and carbon molecules. In addition to this long established metabolic function, GAPDH has recently been implicated in several non-metabolic processes, including transcription activation, initiation of apoptosis, and ER to Golgi vesicle shuttling.
Catalog No. 50-125-2742