Learn More
Aviva Systems Biology Corp GAPDH Antibody - middle region (ARP58578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAPDH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GAPDH. Alternate names: G3PD, GAPD, MGC88685ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-62358