Learn More
Santa Cruz Biotechnology GAPDH ANTIBODY (6C5)
Supplier:  Santa Cruz Biotechnology SC32233
Encompass
GAPDH (6C5) mouse monoclonal IgG1; Raised against GAPDH purified from rabbit muscle; Species: M, R, H; Applications: WB, IP, IF; Storage: 4 deg.C; 200ug/mL
Catalog No. NC9407190