Learn More
Biomatik Corporation Gapdh Recombinant Antibody
Supplier:  Biomatik Corporation RAC07056
Encompass_Preferred
Gapdh Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to GAPDH. The Gapdh Recombinant Antibody has been validated for the following applications: ELISA, WB. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide. Applications: ELISA, WB. Target Synonyms: GAPDH. Accession Number: P04406. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5643