Learn More
Genetex Inc Anti-GAPDH antibody [GT239]
SDP
Supplier:  Genetex Inc GTX627408
Encompass
The Anti-GAPDH antibody [GT239] from GeneTex is a Mouse Monoclonal antibody to GAPDH. The Anti-GAPDH antibody [GT239] Recognizes Human, Mouse, Drosophila, Monkey, Rat, Zebrafish, Yeast, E. coli, Hamster. The Anti-GAPDH antibody [GT239] has been validated for the following Applications: EMSA, ICC/IF, IHC-P, WB.
Catalog No. NC0992758