Learn More
Medchemexpress LLC GAPDH Human HEK293 100ug | 100ug
SDP
Catalog No. 5000194936
Encompass_Preferred
The GAPDH protein has dual roles in glycolysis and nuclear function It also affects the organization and assembly of the cytoskeleton and promotes the association of microtubules and membranes GAPDH Protein Human (HEK293 His) is the recombinant human-derived GAPDH protein expressed by HEK293 with N-6 His labeled tag
Catalog No. 50-001-94936 Supplier Medchemexpress LLC Supplier No. HYP78776100UG
The GAPDH protein has dual roles in glycolysis and nuclear function It also affects the organization and assembly of the cytoskeleton and promotes the association of microtubules and membranes GAPDH Protein Human (HEK293 His) is the recombinant human-derived GAPDH protein expressed by HEK293 with N-6 His labeled tag