Learn More
Medchemexpress LLC GAPDH Human HEK293 5ug | 5ug
SDP
Catalog No. 5000194950
Encompass_Preferred
The GAPDH protein has dual roles in glycolysis and nuclear function It also affects the organization and assembly of the cytoskeleton and promotes the association of microtubules and membranes GAPDH Protein Human (HEK293 His) is the recombinant human-derived GAPDH protein expressed by HEK293 with N-6 His labeled tag
Catalog No. 50-001-94950 Supplier Medchemexpress LLC Supplier No. HYP787765UG
The GAPDH protein has dual roles in glycolysis and nuclear function It also affects the organization and assembly of the cytoskeleton and promotes the association of microtubules and membranes GAPDH Protein Human (HEK293 His) is the recombinant human-derived GAPDH protein expressed by HEK293 with N-6 His labeled tag