Learn More
Cayman Chemical GAPDH InhIbItr Compound F8 1mg
SDP
Catalog No. 502945159
Encompass_Preferred
A covalent inhibitor of GAPDH (IC50 = 39.31 µM); decreases viability of, and glucose metabolism in, HEK293 cells in a concentration-dependent manner
Catalog No. 50-294-5159 Supplier Cayman Chemical Supplier No. 420901 MG
A covalent inhibitor of GAPDH (IC50 = 39.31 µM); decreases viability of, and glucose metabolism in, HEK293 cells in a concentration-dependent manner